siRNA Details
| Gene Symbol: Tmem159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194354.2 | NP_919335.2 | 6 | 625 | ||
| XM_006230138.2 | XP_006230200.1 | 5 | 718 | |||
| XM_006230139.2 | XP_006230201.1 | 6 | 760 | |||
| XM_008759784.1 | XP_008758006.1 | 5 | 1556 | |||
| XM_008759785.1 | XP_008758007.1 | 5 | 1551 | |||
| GenBank mRNA | AY368496.1 | - | 4 | 459 | ||
| BC076384.1 | - | 6 | 625 | 7107584 | ||

Tmem159
Genomic Map