siRNA Details
| Gene Symbol: Spire1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176832.2 | NP_789802.1 | 11 | 1233 | ||
| NM_194355.2 | NP_919336.1 | 12 | 1505 | |||
| GenBank mRNA | AK145500.1 | - | 1 | 965 | ||
| BC046486.1 | - | 5 | 588 | |||
| BC058669.1 | - | 12 | 1373 | |||
| BC079633.1 | - | 2 | 81 | |||
| BC094375.1 | - | 12 | 1426 | 30535423 | ||
| BQ178805.1 | - | 4 | 398 | |||

Spire1
Genomic Map