siRNA Details
| Gene Symbol: Spire1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176832.2 | NP_789802.1 | 14 | 3694 | ||
| NM_194355.2 | NP_919336.1 | 15 | 3966 | |||
| GenBank mRNA | AK044274.1 | - | 1 | 987 | ||
| AK049885.1 | - | 1 | 1455 | |||
| AK049922.1 | - | 1 | 672 | |||
| AK082780.1 | - | 1 | 216 | |||
| BC046486.1 | - | 8 | 3049 | |||
| BC058669.1 | - | 15 | 3834 | |||
| BC094375.1 | - | 15 | 3887 | 30535423 | ||

Spire1
Genomic Map