siRNA Details
| Gene Symbol: Cdk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194444.2 | NP_919426.2 | 11 | 851 | ||
| NM_194446.2 | NP_919428.1 | 11 | 889 | |||
| GenBank mRNA | AK133918.1 | - | 11 | 844 | ||
| AK144801.1 | - | 11 | 861 | |||
| AK170594.1 | - | 11 | 885 | |||
| BC017131.1 | - | 11 | 826 | 4506458 | ||
| BC023736.1 | - | 11 | 870 | |||
| BC052096.1 | - | 8 | 1095 | |||
| DQ288857.1 | - | 11 | 888 | |||

Cdk10
Genomic Map