siRNA Details
| Gene Symbol: Ankrd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030886.2 | NP_112148.2 | 24 | 4478 | ||
| NM_198010.2 | NP_932127.2 | 23 | 3725 | |||
| GenBank mRNA | AF130371.1 | - | 22 | 3266 | ||
| AK153560.1 | - | 3 | 233 | |||
| AK160263.1 | - | 9 | 1242 | |||
| BC080807.1 | - | 14 | 1748 | |||
| BC085166.1 | - | 23 | 3243 | |||

Ankrd17
Genomic Map