siRNA Details
| Gene Symbol: Trmt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198051.1 | NP_932168.1 | 11 | 1086 | ||
| XM_006227748.2 | XP_006227810.1 | 11 | 1100 | |||
| XM_006227749.2 | XP_006227811.1 | 11 | 1617 | |||
| XM_006227750.2 | XP_006227812.1 | 11 | 1099 | |||
| XM_006227751.2 | XP_006227813.1 | 11 | 1098 | |||
| XM_006227752.2 | XP_006227814.1 | 11 | 1098 | |||
| XM_008758673.1 | XP_008756895.1 | 11 | 1760 | |||
| GenBank mRNA | AF532978.1 | - | 11 | 1086 | ||

Trmt11
Genomic Map