siRNA Details
Gene Symbol: Qrfpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198199.1 | NP_937842.1 | 5 | 815 | ||
GenBank mRNA | AB109630.1 | - | 5 | 815 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
