siRNA Details
| Gene Symbol: TBC1D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015079.5 | NP_055894.6 | 9 | 1807 | ||
| NM_144572.1 | NP_653173.1 | 9 | 1807 | |||
| XM_011521387.2 | XP_011519689.1 | 8 | 1752 | |||
| XM_017022022.1 | XP_016877511.1 | 11 | 1828 | |||
| GenBank mRNA | AB028978.1 | - | 9 | 1670 | ||
| AK000173.1 | - | 2 | 774 | |||
| BC033712.1 | - | 4 | 590 | |||
| BC140766.1 | - | 8 | 1452 | 9021634 | ||
| BC144482.1 | - | 8 | 1452 | 9053009 | ||
| BC152465.1 | - | 9 | 1670 | 8862601 | ||

TBC1D2B
Genomic Map