siRNA Details
| Gene Symbol: Ly6g5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198739.3 | NP_942034.3 | 2 | 167 | ||
| XM_008772716.1 | XP_008770938.1 | 2 | 168 | |||
| GenBank mRNA | AJ515240.1 | - | 2 | 167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6g5c| Gene Symbol: Ly6g5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198739.3 | NP_942034.3 | 2 | 167 | ||
| XM_008772716.1 | XP_008770938.1 | 2 | 168 | |||
| GenBank mRNA | AJ515240.1 | - | 2 | 167 | ||