siRNA Details
| Gene Symbol: Tpd52l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198744.2 | NP_942039.1 | 8 | 1287 | ||
| XM_006235754.2 | XP_006235816.1 | 9 | 1313 | |||
| XM_006235757.2 | XP_006235819.1 | 8 | 1273 | |||
| XM_006235759.2 | XP_006235821.1 | 7 | 1237 | |||
| XM_006235760.2 | XP_006235822.1 | 7 | 1241 | |||
| XM_006235761.2 | XP_006235823.1 | 6 | 1182 | |||
| XM_008762499.1 | XP_008760721.1 | 9 | 1247 | |||
| XM_008762500.1 | XP_008760722.1 | 6 | 1116 | |||
| GenBank mRNA | AF329826.1 | - | 1 | 197 | ||
| BC059167.1 | - | 8 | 1288 | 6890828 | ||

Tpd52l2
Genomic Map