siRNA Details
| Gene Symbol: Tmem110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198774.2 | NP_942069.1 | 8 | 1389 | ||
| GenBank mRNA | AY283801.1 | - | 8 | 1412 | ||
| BC088092.1 | - | 8 | 1389 | 7368117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem110| Gene Symbol: Tmem110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198774.2 | NP_942069.1 | 8 | 1389 | ||
| GenBank mRNA | AY283801.1 | - | 8 | 1412 | ||
| BC088092.1 | - | 8 | 1389 | 7368117 | ||