siRNA Details
| Gene Symbol: Lamtor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199102.1 | NP_954533.1 | 5 | 941 | ||
| GenBank mRNA | BC061783.1 | - | 5 | 941 | 5623933 | |
| FQ214531.1 | - | 5 | 945 | |||
| FQ219988.1 | - | 5 | 954 | |||
| FQ220581.1 | - | 5 | 937 | |||
| FQ220693.1 | - | 5 | 939 | |||
| Gene Symbol: LOC100361543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730236.3 | XP_002730282.1 | 1 | 918 | ||

Lamtor1
Genomic Map