siRNA Details
| Gene Symbol: Cnppd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199112.1 | NP_954543.1 | 9 | 1114 | ||
| XM_006245226.2 | XP_006245288.1 | 9 | 1148 | |||
| GenBank mRNA | BC061750.1 | - | 9 | 1114 | 5599704 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnppd1| Gene Symbol: Cnppd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199112.1 | NP_954543.1 | 9 | 1114 | ||
| XM_006245226.2 | XP_006245288.1 | 9 | 1148 | |||
| GenBank mRNA | BC061750.1 | - | 9 | 1114 | 5599704 | |