siRNA Details
| Gene Symbol: BC051628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199312.3 | NP_955016.1 | 3 | 1291 | ||
| GenBank mRNA | AK030149.1 | - | 3652 | |||
| BC051628.1 | - | 3 | 1291 | 6742098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC051628| Gene Symbol: BC051628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199312.3 | NP_955016.1 | 3 | 1291 | ||
| GenBank mRNA | AK030149.1 | - | 3652 | |||
| BC051628.1 | - | 3 | 1291 | 6742098 | ||