siRNA Details
Gene Symbol: Cd200r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206535.1 | NP_996258.1 | 6 | 1118 | ||
GenBank mRNA | AY230198.1 | - | 6 | 1118 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd200r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206535.1 | NP_996258.1 | 6 | 1118 | ||
GenBank mRNA | AY230198.1 | - | 6 | 1118 |