siRNA Details
| Gene Symbol: ZZEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015113.3 | NP_055928.3 | 32 | 5205 | ||
| XM_005256561.2 | XP_005256618.1 | 32 | 5269 | |||
| XM_011523759.2 | XP_011522061.1 | 32 | 5269 | |||
| XM_017024382.1 | XP_016879871.1 | 32 | 5269 | |||
| XM_017024383.1 | XP_016879872.1 | 32 | 5266 | |||
| XM_017024384.1 | XP_016879873.1 | 7 | 1080 | |||
| GenBank mRNA | AB007859.4 | - | 32 | 5205 | ||
| AK001683.1 | - | 3 | 304 | |||
| AK122719.1 | - | 7 | 1299 | |||
| BC047285.1 | - | 4 | 597 | |||
| BC151836.1 | - | 32 | 5205 | 8860131 | ||

ZZEF1
Genomic Map