siRNA Details
| Gene Symbol: GPR171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013308.3 | NP_037440.3 | 3 | 758 | ||
| XM_005247402.3 | XP_005247459.1 | 2 | 1142 | |||
| XM_005247403.3 | XP_005247460.1 | 2 | 793 | |||
| XM_017006274.1 | XP_016861763.1 | 3 | 1235 | |||
| GenBank mRNA | AF002986.1 | - | 3 | 737 | ||
| BC036815.1 | - | 3 | 749 | 5747378 | ||
More Information
Set Membership: |
|

GPR171
Genomic Map