siRNA Details
| Gene Symbol: UBOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267584.1 | NP_001254513.1 | 3 | 665 | ||
| NM_014948.3 | NP_055763.1 | 3 | 665 | |||
| NM_199415.2 | NP_955447.1 | 3 | 665 | |||
| GenBank mRNA | AB020667.1 | - | 3 | 633 | ||
| AK022444.1 | - | 3 | 629 | |||
| AK222570.1 | - | 3 | 637 | |||
| AK222876.1 | - | 3 | 590 | |||
| BC000515.2 | - | 3 | 585 | 2822816 | ||
| BC046122.1 | - | 3 | 625 | 5554044 | ||
| CR457089.1 | - | 2 | 479 | |||

UBOX5
Genomic Map