siRNA Details
| Gene Symbol: ACIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164814.1 | NP_001158286.1 | 4 | 846 | ||
| NM_014977.3 | NP_055792.1 | 4 | 846 | |||
| XM_005267415.3 | XP_005267472.1 | 4 | 650 | |||
| XM_005267416.3 | XP_005267473.1 | 4 | 650 | |||
| XM_006720081.3 | XP_006720144.1 | 4 | 650 | |||
| XM_011536570.2 | XP_011534872.1 | 4 | 650 | |||
| GenBank mRNA | AB014570.2 | - | 4 | 355 | ||
| AF124726.1 | - | 4 | 845 | |||
| BC140805.1 | - | 4 | 615 | 9021673 | ||

ACIN1
Genomic Map