siRNA Details
| Gene Symbol: POP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145860.1 | NP_001139332.1 | 3 | 343 | ||
| NM_001145861.1 | NP_001139333.1 | 3 | 283 | |||
| NM_015029.2 | NP_055844.2 | 3 | 271 | |||
| XM_011516801.2 | XP_011515103.1 | 3 | 343 | |||
| GenBank mRNA | AK291434.1 | - | 3 | 340 | ||
| BC011529.1 | - | 3 | 271 | 3853321 | ||
| BP249298.1 | - | 3 | 269 | |||
| BP280616.1 | - | 3 | 343 | |||
| X99302.1 | - | 3 | 343 | |||

POP1
Genomic Map