siRNA Details
| Gene Symbol: CMTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015050.2 | NP_055865.1 | 4 | 568 | ||
| XM_005248955.3 | XP_005249012.1 | 4 | 498 | |||
| GenBank mRNA | AK000348.1 | - | 546 | |||
| AK292564.1 | - | 4 | 422 | |||
| BC031890.1 | - | 4 | 462 | 4944457 | ||
| D43949.2 | - | 4 | 458 | |||

CMTR1
Genomic Map