siRNA Details
| Gene Symbol: TRIM35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171982.4 | NP_741983.2 | 2 | 538 | ||
| XM_005273452.3 | XP_005273509.1 | 2 | 533 | |||
| XM_011544453.1 | XP_011542755.1 | 3 | 566 | |||
| GenBank mRNA | AB029021.1 | - | 2 | 488 | ||
| AF492463.1 | - | 2 | 536 | |||
| BC018337.1 | - | 3 | 648 | |||
| BC069226.1 | - | 2 | 480 | 6428667 | ||

TRIM35
Genomic Map