siRNA Details
| Gene Symbol: MON1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286639.1 | NP_001273568.1 | 5 | 2426 | ||
| NM_001286640.1 | NP_001273569.1 | 4 | 2268 | |||
| NM_014940.3 | NP_055755.1 | 6 | 2753 | |||
| GenBank mRNA | AB020679.1 | - | 6 | 2437 | ||
| AK023374.1 | - | 5 | 1694 | |||
| AK128411.1 | - | 3 | 2295 | |||

MON1B
Genomic Map