siRNA Details
| Gene Symbol: ZBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014950.2 | NP_055765.2 | 3 | 3013 | ||
| XM_011536568.2 | XP_011534870.1 | 5 | 3149 | |||
| GenBank mRNA | AB023214.1 | - | 3 | 2884 | ||
| BX537646.1 | - | 1 | 1400 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBTB1| Gene Symbol: ZBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014950.2 | NP_055765.2 | 3 | 3013 | ||
| XM_011536568.2 | XP_011534870.1 | 5 | 3149 | |||
| GenBank mRNA | AB023214.1 | - | 3 | 2884 | ||
| BX537646.1 | - | 1 | 1400 | |||