siRNA Details
| Gene Symbol: SPEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015001.2 | NP_055816.2 | 5 | 1404 | ||
| GenBank mRNA | AF356524.1 | - | 5 | 1404 | ||
| AK127577.1 | - | 3 | 727 | |||
| AL096858.1 | - | 2 | 322 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPEN| Gene Symbol: SPEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015001.2 | NP_055816.2 | 5 | 1404 | ||
| GenBank mRNA | AF356524.1 | - | 5 | 1404 | ||
| AK127577.1 | - | 3 | 727 | |||
| AL096858.1 | - | 2 | 322 | |||