siRNA Details
| Gene Symbol: GGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015044.4 | NP_055859.1 | 5 | 476 | ||
| XM_017023074.1 | XP_016878563.1 | 6 | 4165 | |||
| XM_017023075.1 | XP_016878564.1 | 5 | 479 | |||
| GenBank mRNA | AB029003.1 | - | 2 | 106 | ||
| AF165531.1 | - | 5 | 414 | |||
| AF190863.1 | - | 5 | 420 | |||
| AF233522.1 | - | 5 | 419 | |||
| AF323754.1 | - | 5 | 418 | |||
| AK308425.1 | - | 5 | 463 | |||
| BC000284.1 | - | 5 | 458 | 3356490 | ||
| DA439738.1 | - | 5 | 476 | |||

GGA2
Genomic Map