siRNA Details
| Gene Symbol: RTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015138.4 | NP_055953.3 | 4, 5 | 668 | ||
| GenBank mRNA | AK292302.1 | - | 4, 5 | 640 | ||
| BC015052.2 | - | 4, 5 | 618 | 3909737 | ||
| D87440.2 | - | 4, 5 | 634 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_030112, AK019305, BC117849, BC117848, BC048535, and others.

RTF1
Genomic Map