siRNA Details
| Gene Symbol: SASH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015278.3 | NP_056093.3 | 9 | 1242 | ||
| XM_005266875.1 | XP_005266932.1 | 2 | 468 | |||
| XM_011535640.1 | XP_011533942.1 | 9 | 685 | |||
| XM_017010598.1 | XP_016866087.1 | 9 | 1474 | |||
| XM_017010599.1 | XP_016866088.1 | 9 | 797 | |||
| XM_017010600.1 | XP_016866089.1 | 9 | 685 | |||
| XM_017010601.1 | XP_016866090.1 | 8 | 899 | |||
| XM_017010602.1 | XP_016866091.1 | 7 | 596 | |||
| XM_017010603.1 | XP_016866092.1 | 7 | 535 | |||
| XM_017010604.1 | XP_016866093.1 | 6 | 444 | |||
| XM_017010605.1 | XP_016866094.1 | 6 | 410 | |||
| XM_017010606.1 | XP_016866095.1 | 2 | 468 | |||
| XM_017010607.1 | XP_016866096.1 | 10 | 865 | |||
| GenBank mRNA | AB018333.1 | - | 9 | 985 | ||
| AJ507735.1 | - | 9 | 773 | |||
| AK024403.1 | - | 3 | 227 | |||
| AK025495.1 | - | 9 | 1242 | |||
| AK300403.1 | - | 2 | 152 | |||
| AY927559.1 | - | 2 | 88 | |||
| BC063279.1 | - | 9 | 711 | |||

SASH1
Genomic Map