siRNA Details
| Gene Symbol: WDR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015285.2 | NP_056100.2 | 2 | 328 | ||
| NM_052834.2 | NP_443066.2 | 2 | 328 | |||
| XM_006722431.2 | XP_006722494.1 | 2 | 260 | |||
| XM_011525888.1 | XP_011524190.1 | 2 | 370 | |||
| XM_017025682.1 | XP_016881171.1 | 2 | 233 | |||
| GenBank mRNA | AB011113.2 | - | 2 | 291 | ||
| AF188125.1 | - | 1 | 117 | |||
| AK057141.1 | - | 2 | 352 | |||
| AY099325.1 | - | 1 | 117 | |||
| BC131558.1 | - | 1 | 117 | 40108655 | ||
| BC150282.1 | - | 2 | 291 | 9007268 | ||

WDR7
Genomic Map