siRNA Details
| Gene Symbol: COMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000754.3 | NP_000745.1 | 4 | 661 | ||
| NM_001135161.1 | NP_001128633.1 | 4 | 619 | |||
| NM_001135162.1 | NP_001128634.1 | 4 | 636 | |||
| NM_007310.2 | NP_009294.1 | 2 | 392 | |||
| XM_011529886.1 | XP_011528188.1 | 4 | 566 | |||
| XM_017028594.1 | XP_016884083.1 | 4 | 885 | |||
| XM_017028595.1 | XP_016884084.1 | 4 | 829 | |||
| GenBank mRNA | AK290440.1 | - | 4 | 599 | ||
| BC000419.2 | - | 4 | 589 | 2964400 | ||
| BC005867.2 | - | 4 | 589 | 2964400 | ||
| BX461394.2 | - | 4 | 668 | |||
| CR456422.1 | - | 3 | 427 | |||
| CR456997.1 | - | 2 | 412 | |||
| EF103918.1 | - | 2 | 153 | |||
| EF103922.1 | - | 3 | 441 | |||
| FJ224345.1 | - | 4 | 589 | |||
| FJ410131.1 | - | 2 | 262 | |||
| FJ410132.1 | - | 2 | 412 | |||
| M58525.1 | - | 4 | 616 | |||
| M65212.1 | - | 3 | 490 | |||

COMT
Genomic Map