siRNA Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000777.4 | NP_000768.1 | 11 | 1309 | ||
| NM_001291829.1 | NP_001278758.1 | 12 | 1809 | |||
| NM_001291830.1 | NP_001278759.1 | 12 | 1569 | |||
| NR_033807.2 | 11 | 2913 | ||||
| XM_011515843.1 | XP_011514145.1 | 12 | 1426 | |||
| XM_017011781.1 | XP_016867270.1 | 8 | 951 | |||
| XM_017011782.1 | XP_016867271.1 | 6 | 748 | |||
| GenBank mRNA | AK223008.1 | - | 11 | 1279 | ||
| AK296205.1 | - | 12 | 1536 | |||
| AK313813.1 | - | 11 | 1276 | |||
| BC033862.1 | - | 11 | 1268 | 5188554 | ||
| BX537676.1 | - | 11 | 2884 | |||
| J04813.1 | - | 11 | 1266 | |||

CYP3A5
Genomic Map