siRNA Details
| Gene Symbol: FBXO28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136115.2 | NP_001129587.1 | 4 | 1204 | ||
| NM_015176.3 | NP_055991.1 | 5 | 1400 | |||
| NR_049764.1 | 4 | 1261 | ||||
| GenBank mRNA | AB007952.1 | - | 5 | 1151 | ||
| AK001628.1 | - | 5 | 1376 | |||
| AK303381.1 | - | 4 | 1229 | |||
| AK307892.1 | - | 4 | 1176 | |||
| BC031576.1 | - | 5 | 1369 | 5188219 | ||

FBXO28
Genomic Map