siRNA Details
| Gene Symbol: FAM208A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112736.1 | NP_001106207.1 | 14 | 2143 | ||
| NM_015224.3 | NP_056039.2 | 7 | 1494 | |||
| XM_005264999.1 | XP_005265056.1 | 14 | 2278 | |||
| XM_006713077.2 | XP_006713140.1 | 14 | 2278 | |||
| XM_006713078.2 | XP_006713141.1 | 8 | 1132 | |||
| XM_011533552.2 | XP_011531854.1 | 14 | 2278 | |||
| XM_011533553.2 | XP_011531855.1 | 13 | 1699 | |||
| XM_017006030.1 | XP_016861519.1 | 14 | 2053 | |||
| XM_017006031.1 | XP_016861520.1 | 14 | 2278 | |||
| XM_017006032.1 | XP_016861521.1 | 13 | 1699 | |||
| GenBank mRNA | AF180425.2 | - | 7 | 1778 | ||
| AF273053.1 | - | 1 | 478 | |||
| AK307599.1 | - | 6 | 956 | |||
| BC070096.1 | - | 14 | 2141 | |||
| BC129987.1 | - | 6 | 919 | 40129237 | ||
| BX648955.1 | - | 13 | 1625 | |||

FAM208A
Genomic Map