siRNA Details
| Gene Symbol: SASH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015278.3 | NP_056093.3 | 20 | 4413 | ||
| XM_005266875.1 | XP_005266932.1 | 13 | 3639 | |||
| XM_011535640.1 | XP_011533942.1 | 20 | 3856 | |||
| XM_017010598.1 | XP_016866087.1 | 20 | 5134 | |||
| XM_017010599.1 | XP_016866088.1 | 20 | 4457 | |||
| XM_017010600.1 | XP_016866089.1 | 20 | 4345 | |||
| XM_017010601.1 | XP_016866090.1 | 19 | 4559 | |||
| XM_017010602.1 | XP_016866091.1 | 18 | 4256 | |||
| XM_017010603.1 | XP_016866092.1 | 18 | 4195 | |||
| XM_017010604.1 | XP_016866093.1 | 17 | 4104 | |||
| XM_017010605.1 | XP_016866094.1 | 17 | 4070 | |||
| XM_017010606.1 | XP_016866095.1 | 13 | 4128 | |||
| XM_017010607.1 | XP_016866096.1 | 21 | 4036 | |||
| GenBank mRNA | AB018333.1 | - | 20 | 4156 | ||
| AJ507735.1 | - | 20 | 3944 | |||
| AK024403.1 | - | 14 | 3398 | |||
| AK025495.1 | - | 20 | 4413 | |||
| AK074249.1 | - | 10 | 2698 | |||
| BC028303.1 | - | 3 | 924 | |||
| BC063279.1 | - | 20 | 3882 | |||
| BX537611.1 | - | 9 | 3471 | |||

SASH1
Genomic Map