siRNA Details
| Gene Symbol: FNBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318339.1 | NP_001305268.1 | 7 | 1320 | ||
| NM_015308.3 | NP_056123.2 | 7 | 1314 | |||
| GenBank mRNA | AB023231.2 | - | 7 | 1313 | ||
| AK054636.1 | - | 7 | 1137 | |||
| AK123898.1 | - | 6 | 981 | |||
| AL137480.1 | - | 7 | 1113 | |||
| BC037404.1 | - | 7 | 1145 | 5140963 | ||

FNBP4
Genomic Map