siRNA Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015344.2 | NP_056159.2 | 4 | 1222 | ||
| GenBank mRNA | AF063605.1 | - | 4 | 1199 | ||
| AK075363.1 | - | 4 | 1166 | |||
| AY358674.1 | - | 4 | 1194 | |||
| BC000642.2 | - | 4 | 1205 | 3344487 | ||

LEPROTL1
Genomic Map