siRNA Details
| Gene Symbol: DCAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015397.3 | NP_056212.1 | 6 | 1190 | ||
| XM_011517843.1 | XP_011516145.1 | 6 | 1017 | |||
| GenBank mRNA | AB067479.1 | - | 6 | 1110 | ||
| AF131771.1 | - | 1 | 50 | |||
| AK000354.1 | - | 6 | 968 | |||
| AK292935.1 | - | 6 | 996 | |||
| AL117646.1 | - | 3 | 324 | |||
| BC008893.2 | - | 5 | 753 | |||
| BC063823.1 | - | 6 | 1108 | 6184796 | ||
| DQ518909.1 | - | 6 | 1110 | |||

DCAF12
Genomic Map