siRNA Details
| Gene Symbol: RNF19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001280539.1 | NP_001267468.1 | 3 | 468 | ||
| NM_015435.4 | NP_056250.3 | 3 | 499 | |||
| NM_183419.3 | NP_904355.1 | 2 | 575 | |||
| XM_005250853.3 | XP_005250910.1 | 2 | 537 | |||
| XM_011516956.2 | XP_011515258.1 | 2 | 560 | |||
| XM_017013302.1 | XP_016868791.1 | 3 | 660 | |||
| XM_017013303.1 | XP_016868792.1 | 3 | 570 | |||
| XM_017013304.1 | XP_016868793.1 | 2 | 517 | |||
| XM_017013305.1 | XP_016868794.1 | 1 | 264 | |||
| GenBank mRNA | AB029316.1 | - | 3 | 499 | ||
| AB271913.1 | - | 2 | 346 | |||
| AB271914.1 | - | 3 | 231 | |||
| BC093938.1 | - | 1 | 271 | 7939783 | ||
| BC093940.2 | - | 1 | 271 | 7939785 | ||
| DB037264.1 | - | 2 | 561 | |||
| DB061830.1 | - | 3 | 468 | |||

RNF19A
Genomic Map