siRNA Details
| Gene Symbol: CCDC28A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015439.2 | NP_056254.1 | 1 | 234 | ||
| XM_011535728.2 | XP_011534030.1 | 1 | 237 | |||
| GenBank mRNA | AY167571.1 | - | 1 | 252 | ||
| BC108717.1 | - | 1 | 157 | 6068596 | ||

CCDC28A
Genomic Map