siRNA Details
| Gene Symbol: THUMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114092.1 | NP_001107564.1 | 2, 3 | 408 | ||
| NM_015453.2 | NP_056268.2 | 2, 3 | 613 | |||
| XM_005265021.4 | XP_005265078.1 | 2, 3 | 970 | |||
| XM_005265022.4 | XP_005265079.1 | 2, 3 | 1173 | |||
| XM_005265023.3 | XP_005265080.1 | 2, 3 | 556 | |||
| XM_005265024.4 | XP_005265081.1 | 2, 3 | 556 | |||
| XM_006713086.3 | XP_006713149.1 | 2, 3 | 1212 | |||
| XM_011533570.2 | XP_011531872.1 | 2, 3 | 676 | |||
| XM_011533571.2 | XP_011531873.1 | 2, 3 | 613 | |||
| XM_017006113.1 | XP_016861602.1 | 2, 3 | 970 | |||
| XM_017006114.1 | XP_016861603.1 | 2, 3 | 1212 | |||
| XM_017006115.1 | XP_016861604.1 | 2, 3 | 613 | |||
| XM_017006116.1 | XP_016861605.1 | 2, 3 | 599 | |||
| XM_017006117.1 | XP_016861606.1 | 2, 3 | 970 | |||
| XM_017006118.1 | XP_016861607.1 | 2, 3 | 1212 | |||
| XM_017006119.1 | XP_016861608.1 | 2, 3 | 599 | |||
| GenBank mRNA | AK001680.1 | - | 2, 3 | 372 | ||
| AK023263.1 | - | 2, 3 | 594 | |||
| BC001622.1 | - | 2, 3 | 387 | 3355978 | ||
| BC010421.1 | - | 2, 3 | 944 | 3140059 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008188, U83176, BC002024, AK017246, AK142140, and others.

THUMPD3
Genomic Map