siRNA Details
| Gene Symbol: IBTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300906.1 | NP_001287835.1 | 26 | 4174 | ||
| NM_015525.3 | NP_056340.2 | 26 | 4219 | |||
| XM_006715453.2 | XP_006715516.1 | 26 | 4759 | |||
| XM_006715454.2 | XP_006715517.1 | 26 | 4714 | |||
| GenBank mRNA | AB037838.2 | - | 25 | 3943 | ||
| AF235049.1 | - | 3 | 638 | |||
| AJ420588.1 | - | 1 | 103 | |||
| AJ420593.1 | - | 2 | 123 | |||
| AK126777.1 | - | 10 | 2084 | |||
| AL050018.1 | - | 6 | 759 | |||
| BC027490.1 | - | 4 | 403 | |||
| BC038244.2 | - | 9 | 1057 | |||
| BC042171.1 | - | 5 | 602 | |||
| BC113696.1 | - | 25 | 3748 | 8322748 | ||
| BC113698.1 | - | 25 | 3748 | 8322750 | ||
| BC143736.1 | - | 24 | 3145 | 9052250 | ||
| BC143745.1 | - | 25 | 3704 | 9052259 | ||
| BC143748.1 | - | 23 | 3715 | |||
| DQ005633.1 | - | 26 | 4219 | |||
| DQ005635.1 | - | 2 | 749 | |||

IBTK
Genomic Map