siRNA Details
| Gene Symbol: OSBPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015550.3 | NP_056365.1 | 2, 3 | 569 | ||
| NM_145320.2 | NP_663160.1 | 1, 2 | 242 | |||
| NM_145321.2 | NP_663161.1 | 1, 2 | 242 | |||
| NM_145322.2 | NP_663162.1 | 1, 2 | 242 | |||
| NR_104111.1 | 1, 2 | 242 | ||||
| NR_104112.1 | 1, 2 | 242 | ||||
| XM_005249698.3 | XP_005249755.1 | 2, 3 | 289 | |||
| XM_006715681.3 | XP_006715744.1 | 2, 3 | 556 | |||
| XM_006715682.3 | XP_006715745.1 | 2, 3 | 575 | |||
| XM_006715683.3 | XP_006715746.1 | 2, 3 | 575 | |||
| XM_006715684.3 | XP_006715747.1 | 1, 2 | 242 | |||
| XM_011515258.2 | XP_011513560.1 | 2, 3 | 366 | |||
| GenBank mRNA | AB014604.2 | - | 2, 3 | 506 | ||
| AF323727.1 | - | 2, 3 | 364 | |||
| AF392444.1 | - | 2, 3 | 478 | |||
| AF491781.1 | - | 1, 2 | 196 | |||
| AF491782.1 | - | 1, 2 | 196 | |||
| AF491783.1 | - | 1, 2 | 196 | |||
| AF491784.1 | - | 1, 2 | 196 | |||
| AF491785.1 | - | 1, 2 | 196 | |||
| AF491786.1 | - | 1, 2 | 196 | |||
| AY008372.1 | - | 2, 3 | 399 | |||
| BC017731.1 | - | 2, 3 | 498 | 3909164 | ||

OSBPL3
Genomic Map