siRNA Details
| Gene Symbol: ARHGEF26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251962.1 | NP_001238891.1 | 2 | 772 | ||
| NM_001251963.1 | NP_001238892.1 | 2 | 699 | |||
| NM_015595.3 | NP_056410.3 | 2 | 699 | |||
| XM_011512672.1 | XP_011510974.1 | 2 | 699 | |||
| GenBank mRNA | AB073386.1 | - | 2 | 619 | ||
| AF415175.1 | - | 2 | 654 | |||
| AK123040.1 | - | 2 | 643 | |||
| AK127282.1 | - | 2 | 772 | |||
| AY552599.1 | - | 1 | 488 | |||
| BC078655.1 | - | 2 | 640 | 4374088 | ||

ARHGEF26
Genomic Map