siRNA Details
| Gene Symbol: TBC1D29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015594.2 | NP_056409.1 | 3, 4 | 333 | ||
| XM_011524596.1 | XP_011522898.1 | 3, 4 | 259 | |||
| GenBank mRNA | AL117394.1 | - | 4, 5 | 2682 | ||
| BC096718.1 | - | 3, 4 | 333 | 40003088 | ||

TBC1D29
Genomic Map