siRNA Details
| Gene Symbol: SPEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK313766.1 | - | 4 | 612 | ||
| AL080154.1 | - | 4 | 581 | |||
| BC022476.1 | - | 4 | 558 | 4816764 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPEF1| Gene Symbol: SPEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK313766.1 | - | 4 | 612 | ||
| AL080154.1 | - | 4 | 581 | |||
| BC022476.1 | - | 4 | 558 | 4816764 | ||