siRNA Details
| Gene Symbol: WWTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168278.1 | NP_001161750.1 | 8 | 1725 | ||
| NM_001168280.1 | NP_001161752.1 | 7 | 1672 | |||
| NM_015472.4 | NP_056287.1 | 7 | 1808 | |||
| XM_017006121.1 | XP_016861610.1 | 10 | 1918 | |||
| XM_017006122.1 | XP_016861611.1 | 8 | 1717 | |||
| GenBank mRNA | AK130514.1 | - | 1 | 69 | ||
| AL050107.1 | - | 5 | 1032 | |||
| AL833852.1 | - | 6 | 1462 | |||
| BC014052.2 | - | 7 | 1702 | 3635491 | ||

WWTR1
Genomic Map