siRNA Details
| Gene Symbol: SH2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF227967.1 | - | 2778 | |||
| AF227968.1 | - | 2878 | ||||
| AF227969.1 | - | 2931 | ||||
| AL049924.1 | - | 2375 | ||||
| AL713760.1 | - | 4852 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH2B1| Gene Symbol: SH2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF227967.1 | - | 2778 | |||
| AF227968.1 | - | 2878 | ||||
| AF227969.1 | - | 2931 | ||||
| AL049924.1 | - | 2375 | ||||
| AL713760.1 | - | 4852 | ||||