siRNA Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114600.2 | NP_001108072.1 | 4 | 625 | ||
| NM_001271869.1 | NP_001258798.1 | 4 | 622 | |||
| NR_073500.1 | 2 | 320 | ||||
| NR_073501.1 | 5 | 696 | ||||
| NR_073502.1 | 3 | 575 | ||||
| NR_073503.1 | 3 | 572 | ||||
| GenBank mRNA | AB097042.1 | - | 3 | 363 | ||
| AB097043.1 | - | 3 | 363 | |||
| AB097044.1 | - | 3 | 354 | |||
| AB097045.1 | - | 3 | 354 | |||
| AB097046.1 | - | 4 | 413 | |||
| AJ295987.1 | - | 3 | 359 | |||
| AK297926.1 | - | 4 | 413 | |||
| AK308787.1 | - | 2 | 107 | |||
| AL050028.1 | - | 3 | 359 | |||
| BC023988.1 | - | 3 | 380 | 3825796 | ||
| BC065538.1 | - | 3 | 360 | 6053254 | ||
| DA010145.1 | - | 4 | 417 | |||
| DA438597.1 | - | 5 | 483 | |||

SZRD1
Genomic Map