siRNA Details
| Gene Symbol: KIF1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015634.3 | NP_056449.1 | 1 | 210 | ||
| GenBank mRNA | AA913814.1 | - | 1 | 202 | ||
| AB033105.1 | - | 1 | 133 | |||
| AK291343.1 | - | 1 | 115 | |||
| BC006427.1 | - | 1 | 133 | |||
| BC012180.1 | - | 1 | 107 | 4550085 | ||
| DA292451.1 | - | 1 | 213 | |||

KIF1BP
Genomic Map