siRNA Details
| Gene Symbol: RGS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286692.1 | NP_001273621.1 | 4 | 332 | ||
| NM_001286693.1 | NP_001273622.1 | 4 | 446 | |||
| NM_015668.4 | NP_056483.3 | 4 | 332 | |||
| XM_005250856.2 | XP_005250913.1 | 4 | 178 | |||
| XM_005250857.2 | XP_005250914.1 | 4 | 178 | |||
| XM_005250858.2 | XP_005250915.1 | 4 | 178 | |||
| XM_005250860.2 | XP_005250917.1 | 4 | 178 | |||
| XM_006716540.2 | XP_006716603.1 | 4 | 446 | |||
| XM_011516957.1 | XP_011515259.1 | 5 | 627 | |||
| XM_011516958.1 | XP_011515260.1 | 4 | 525 | |||
| XM_011516959.2 | XP_011515261.1 | 4 | 178 | |||
| XM_017013309.1 | XP_016868798.1 | 4 | 178 | |||
| XM_017013310.1 | XP_016868799.1 | 4 | 178 | |||
| GenBank mRNA | AK292559.1 | - | 4 | 178 | ||
| AY009106.1 | - | 4 | 161 | |||
| BC036665.1 | - | 4 | 183 | 5268331 | ||
| BC047060.1 | - | 4 | 216 | |||
| BC092411.1 | - | 4 | 332 | 30386231 | ||

RGS22
Genomic Map